The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003323.2 | NP_003314.2 | 10 - 11 | 1329 | 75 |
XM_006723347.3 | XP_006723410.1 | 10 - 11 | 1329 | 75 | |
XM_006723348.3 | XP_006723411.1 | 10 - 11 | 1273 | 75 | |
XM_011527252.2 | XP_011525554.1 | 11 - 12 | 1433 | 75 | |
XM_011527253.2 | XP_011525555.1 | 10 - 11 | 1320 | 75 | |
XM_011527255.2 | XP_011525557.1 | 9 - 10 | 1277 | 75 | |
XM_011527256.2 | XP_011525558.1 | 8 - 9 | 1244 | 75 | |
GenBank mRNA | BC026070.2 | - | 10 - 11 | 1323 | 75 |
U82469.1 | - | 10 - 11 | 1271 | 75 |