Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TULP2
Entrez Gene ID
7288
Gene Name
tubby like protein 2
Gene Aliases
CT65, TUBL2
Chromosome Location
Chr.19: 48880817 - 48900177 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003323.2 NP_003314.2 1 - 2 153 73
XM_006723347.3 XP_006723410.1 1 - 2 153 73
XM_011527252.2 XP_011525554.1 2 - 3 257 73
XM_011527253.2 XP_011525555.1 1 - 2 153 73
XM_011527254.2 XP_011525556.1 1 - 2 153 73
GenBank mRNA BC026070.2 - 1 - 2 147 73
DB442827.1 - 1 - 2 153 73
U82469.1 - 1 - 2 95 73

View Full Product Details