The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016216.3 | NP_057300.2 | 6 - 7 | 948 | 115 |
GenBank mRNA | AF180919.2 | - | 6 - 7 | 902 | 115 |
AK000116.1 | - | 1 - 2 | 425 | 115 | |
AK094147.1 | - | 7 - 8 | 833 | 115 | |
BC009472.1 | - | 6 - 7 | 876 | 115 |