Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZMYND10
Entrez Gene ID
51364
Gene Name
zinc finger MYND-type containing 10
Gene Aliases
BLU, CILD22, FLU
Chromosome Location
Chr.3: 50341106 - 50346028 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015896.3 NP_056980.2 6 - 7 767 67
XM_005265216.2 XP_005265273.1 5 - 6 940 67
GenBank mRNA AK058135.1 - 6 - 7 697 67
AK096525.1 - 6 - 7 1875 67
AK223343.1 - 6 - 7 682 67
AL833828.1 - 6 - 7 696 67
BC033732.1 - 6 - 7 663 67
U70824.1 - 6 - 7 721 67

View Full Product Details