Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NINL
Entrez Gene ID
22981
Gene Name
ninein like
Gene Aliases
NLP
Chromosome Location
Chr.20: 25452697 - 25585531 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318226.1 NP_001305155.1 13 - 14 1762 79
NM_025176.5 NP_079452.3 13 - 14 1762 79
XM_011529186.2 XP_011527488.1 13 - 14 2400 79
XM_011529187.2 XP_011527489.1 13 - 14 2385 79
XM_011529188.1 XP_011527490.1 13 - 14 1777 79
XM_011529189.1 XP_011527491.1 13 - 14 1785 79
XM_011529190.1 XP_011527492.1 12 - 13 1700 79
XM_011529191.1 XP_011527493.1 9 - 10 1292 79
XM_011529192.1 XP_011527494.1 6 - 7 2212 79
XM_011529193.1 XP_011527495.1 12 - 13 1700 79
XM_011529194.2 XP_011527496.1 4 - 5 1481 79
XM_011529195.2 XP_011527497.1 3 - 4 306 79
XM_011529197.2 XP_011527499.1 5 - 6 675 79
XM_017027732.1 XP_016883221.1 14 - 15 3719 79
XM_017027733.1 XP_016883222.1 14 - 15 1831 79
XM_017027734.1 XP_016883223.1 7 - 8 866 79
XM_017027735.1 XP_016883224.1 3 - 4 311 79
GenBank mRNA AB023197.1 - 13 - 14 1748 79
AK093569.1 - 3 - 4 1145 79
AK309298.1 - 13 - 14 1711 79
BC036380.2 - 12 - 13 1700 79
EU718622.1 - 13 - 14 1748 79

View Full Product Details