Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NINL
Entrez Gene ID
22981
Gene Name
ninein like
Gene Aliases
NLP
Chromosome Location
Chr.20: 25452697 - 25585531 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318226.1 NP_001305155.1 8 - 9 1122 85
NM_025176.5 NP_079452.3 8 - 9 1122 85
XM_011529187.2 XP_011527489.1 8 - 9 1745 85
XM_017027733.1 XP_016883222.1 9 - 10 1191 85
GenBank mRNA AB023197.1 - 8 - 9 1108 85
AK293991.1 - 8 - 9 1408 85
BC036380.2 - 7 - 8 1060 85
EU718622.1 - 8 - 9 1108 85

View Full Product Details