The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206794.1 | NP_001193723.1 | 7 - 8 | 826 | 127 |
NM_016374.5 | NP_057458.4 | 7 - 8 | 947 | 127 | |
NM_031371.3 | NP_112739.2 | 7 - 8 | 947 | 127 | |
NR_135074.1 | 7 - 8 | 949 | 127 | ||
XM_006711781.2 | XP_006711844.1 | 7 - 8 | 970 | 127 | |
XM_011544212.2 | XP_011542514.1 | 7 - 8 | 786 | 127 | |
XM_017001468.1 | XP_016856957.1 | 6 - 7 | 608 | 127 | |
XM_017001469.1 | XP_016856958.1 | 6 - 7 | 609 | 127 | |
XM_017001470.1 | XP_016856959.1 | 6 - 7 | 607 | 127 | |
XM_017001471.1 | XP_016856960.1 | 6 - 7 | 611 | 127 | |
GenBank mRNA | AB210032.1 | - | 7 - 8 | 826 | 127 |
AF208045.2 | - | 6 - 7 | 493 | 127 | |
AF214114.2 | - | 7 - 8 | 913 | 127 | |
AK057552.1 | - | 7 - 8 | 947 | 127 | |
AY220790.1 | - | 5 - 6 | 449 | 127 | |
BC048959.1 | - | 7 - 8 | 908 | 127 | |
BC062536.1 | - | 7 - 8 | 897 | 127 | |
BC104632.1 | - | 5 - 6 | 456 | 127 | |
BC130418.1 | - | 7 - 8 | 864 | 127 | |
BF899710.1 | - | 4 - 5 | 127 | ||
BX648820.1 | - | 7 - 8 | 899 | 127 |