Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KYNU
Entrez Gene ID
8942
Gene Name
kynureninase
Gene Aliases
KYNUU
Chromosome Location
Chr.2: 142877626 - 143042316 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199241.1 NP_001186170.1 11 - 12 1118 79
NM_003937.2 NP_003928.1 10 - 11 1032 79
XM_011512102.2 XP_011510404.1 9 - 10 841 79
GenBank mRNA AK223426.1 - 10 - 11 1030 79
AK292095.1 - 10 - 11 996 79
AK315343.1 - 11 - 12 1080 79
CR457423.1 - 9 - 10 902 79
U57721.1 - 10 - 11 1008 79

View Full Product Details