The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032998.1 | NP_001028170.1 | 8 - 9 | 857 | 127 |
NM_001199241.1 | NP_001186170.1 | 9 - 10 | 943 | 127 | |
NM_003937.2 | NP_003928.1 | 8 - 9 | 857 | 127 | |
XM_011512102.2 | XP_011510404.1 | 7 - 8 | 666 | 127 | |
XM_017005217.1 | XP_016860706.1 | 8 - 9 | 819 | 127 | |
GenBank mRNA | AK223426.1 | - | 8 - 9 | 855 | 127 |
AK290900.1 | - | 8 - 9 | 803 | 127 | |
AK292095.1 | - | 8 - 9 | 821 | 127 | |
AK315343.1 | - | 9 - 10 | 905 | 127 | |
BC000879.1 | - | 8 - 9 | 826 | 127 | |
CR457423.1 | - | 7 - 8 | 727 | 127 | |
U57721.1 | - | 8 - 9 | 833 | 127 |