The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000390.3 | NP_000381.1 | 13 - 14 | 1637 | 81 |
NM_001320959.1 | NP_001307888.1 | 13 - 14 | 1634 | 81 | |
XM_017029242.1 | XP_016884731.1 | 13 - 14 | 1633 | 81 | |
XM_017029243.1 | XP_016884732.1 | 13 - 14 | 1724 | 81 | |
XM_017029244.1 | XP_016884733.1 | 13 - 14 | 1622 | 81 | |
XM_017029245.1 | XP_016884734.1 | 13 - 14 | 1724 | 81 | |
XM_017029246.1 | XP_016884735.1 | 13 - 14 | 1603 | 81 | |
GenBank mRNA | AK291160.1 | - | 13 - 14 | 1626 | 81 |
AK299323.1 | - | 10 - 11 | 1342 | 81 | |
AK312443.1 | - | 13 - 14 | 1626 | 81 | |
M83773.1 | - | 7 - 8 | 953 | 81 | |
X57637.1 | - | 6 - 7 | 719 | 81 | |
X78121.1 | - | 13 - 14 | 1634 | 81 |