The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278270.1 | NP_001265199.1 | 5 - 6 | 924 | 63 |
NM_006354.3 | NP_006345.1 | 5 - 6 | 1530 | 63 | |
NM_133480.2 | NP_597814.1 | 5 - 6 | 1530 | 63 | |
NR_103488.1 | 3 - 4 | 966 | 63 | ||
GenBank mRNA | AF069733.1 | - | 4 - 5 | 707 | 63 |
AK000228.1 | - | 5 - 6 | 934 | 63 | |
AK292264.1 | - | 5 - 6 | 924 | 63 | |
AL117487.1 | - | 5 - 6 | 1193 | 63 | |
BC009240.2 | - | 5 - 6 | 1255 | 63 | |
BC013433.1 | - | 5 - 6 | 1193 | 63 | |
BG477633.1 | - | 4 - 5 | 228 | 63 | |
CR533543.1 | - | 4 - 5 | 707 | 63 |