The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319683.1 | NP_001306612.1 | 2 - 3 | 157 | 101 |
NM_053274.2 | NP_444504.1 | 2 - 3 | 157 | 101 | |
NR_135089.1 | 2 - 3 | 157 | 101 | ||
XM_006710309.2 | XP_006710372.1 | 2 - 3 | 135 | 101 | |
XM_011540546.2 | XP_011538848.1 | 4 - 5 | 320 | 101 | |
XM_017000137.1 | XP_016855626.1 | 2 - 3 | 525 | 101 | |
XM_017000138.1 | XP_016855627.1 | 2 - 3 | 525 | 101 | |
XM_017000139.1 | XP_016855628.1 | 2 - 3 | 525 | 101 | |
XM_017000141.1 | XP_016855630.1 | 2 - 3 | 157 | 101 | |
XM_017000142.1 | XP_016855631.1 | 2 - 3 | 135 | 101 | |
XM_017000143.1 | XP_016855632.1 | 2 - 3 | 516 | 101 | |
XM_017000144.1 | XP_016855633.1 | 2 - 3 | 122 | 101 | |
GenBank mRNA | AJ302735.1 | - | 2 - 3 | 123 | 101 |
AJ347709.1 | - | 2 - 3 | 124 | 101 | |
AK295971.1 | - | 2 - 3 | 135 | 101 | |
BC001257.1 | - | 2 - 3 | 124 | 101 | |
DA729622.1 | - | 2 - 3 | 157 | 101 | |
U73704.1 | - | 2 - 3 | 135 | 101 |