The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319683.1 | NP_001306612.1 | 7 - 8 | 839 | 83 |
NM_053274.2 | NP_444504.1 | 7 - 8 | 839 | 83 | |
NR_135089.1 | 7 - 8 | 839 | 83 | ||
XM_005270401.3 | XP_005270458.1 | 6 - 7 | 701 | 83 | |
XM_006710309.2 | XP_006710372.1 | 7 - 8 | 841 | 83 | |
XM_011540546.2 | XP_011538848.1 | 9 - 10 | 1002 | 83 | |
XM_017000137.1 | XP_016855626.1 | 7 - 8 | 1207 | 83 | |
XM_017000138.1 | XP_016855627.1 | 7 - 8 | 1207 | 83 | |
XM_017000139.1 | XP_016855628.1 | 7 - 8 | 1207 | 83 | |
XM_017000140.1 | XP_016855629.1 | 6 - 7 | 817 | 83 | |
XM_017000141.1 | XP_016855630.1 | 7 - 8 | 839 | 83 | |
XM_017000142.1 | XP_016855631.1 | 7 - 8 | 841 | 83 | |
XM_017000143.1 | XP_016855632.1 | 7 - 8 | 1222 | 83 | |
GenBank mRNA | AJ302735.1 | - | 7 - 8 | 805 | 83 |
AJ347709.1 | - | 7 - 8 | 806 | 83 | |
AK295971.1 | - | 7 - 8 | 817 | 83 | |
BC001257.1 | - | 7 - 8 | 806 | 83 | |
U73704.1 | - | 7 - 8 | 817 | 83 |