The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015174.1 | NP_055989.1 | 17 - 18 | 2564 | 135 |
XM_011527831.2 | XP_011526133.1 | 17 - 18 | 2566 | 135 | |
GenBank mRNA | AB029009.1 | - | 16 - 17 | 2442 | 135 |