The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015174.1 | NP_055989.1 | 3 - 4 | 392 | 86 |
XM_011527831.2 | XP_011526133.1 | 3 - 4 | 391 | 86 | |
GenBank mRNA | AB029009.1 | - | 2 - 3 | 270 | 86 |
AK295037.1 | - | 3 - 4 | 391 | 86 |