The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015174.1 | NP_055989.1 | 9 - 10 | 1505 | 95 |
XM_011527831.2 | XP_011526133.1 | 9 - 10 | 1504 | 95 | |
GenBank mRNA | AB029009.1 | - | 8 - 9 | 1383 | 95 |
AK295037.1 | - | 8 - 9 | 1326 | 95 |