Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
B3GALNT2
Entrez Gene ID
148789
Gene Name
beta-1,3-N-acetylgalactosaminyltransferase 2
Gene Aliases
B3GalNAc-T2, MDDGA11
Chromosome Location
Chr.1: 235440654 - 235504481 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277155.2 NP_001264084.1 6 - 7 997 64
NM_152490.4 NP_689703.1 5 - 6 874 64
XM_006711749.3 XP_006711812.1 5 - 6 816 64
XM_017000394.1 XP_016855883.1 6 - 7 997 64
XM_017000395.1 XP_016855884.1 6 - 7 950 64
GenBank mRNA AB209046.1 - 4 - 5 428 64
AK091497.1 - 4 - 5 546 64
BC016974.1 - 6 - 7 938 64
BC029564.1 - 5 - 6 874 64

View Full Product Details