The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004997.2 | NP_004988.2 | 7 - 8 | 1152 | 145 |
GenBank mRNA | BC044226.1 | - | 7 - 8 | 1152 | 145 |
L05606.1 | - | 7 - 8 | 1121 | 145 |