The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024686.4 | NP_078962.4 | 12 - 13 | 1750 | 120 |
XM_005271208.2 | XP_005271265.1 | 11 - 12 | 1561 | 120 | |
XM_011542159.2 | XP_011540461.1 | 12 - 13 | 1940 | 120 | |
XM_017002347.1 | XP_016857836.1 | 13 - 14 | 1770 | 120 | |
XM_017002348.1 | XP_016857837.1 | 12 - 13 | 1940 | 120 | |
XM_017002349.1 | XP_016857838.1 | 11 - 12 | 1525 | 120 | |
GenBank mRNA | AK026686.1 | - | 5 - 6 | 516 | 120 |
AK094174.1 | - | 7 - 8 | 1352 | 120 | |
AK126792.1 | - | 12 - 13 | 1647 | 120 | |
AK128604.1 | - | 9 - 10 | 1729 | 120 | |
AK225574.1 | - | 5 - 6 | 516 | 120 | |
AY170843.1 | - | 11 - 12 | 1524 | 120 | |
BC028350.2 | - | 5 - 6 | 686 | 120 | |
BC048970.1 | - | 12 - 13 | 1692 | 120 | |
BC060878.1 | - | 12 - 13 | 1732 | 120 |