The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021186.4 | NP_067009.1 | 2 - 3 | 581 | 84 |
GenBank mRNA | AK314151.1 | - | 2 - 3 | 581 | 84 |
BC069521.1 | - | 2 - 3 | 294 | 84 |