The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005957.4 | NP_005948.3 | 2 - 3 | 471 | 79 |
XM_005263458.3 | XP_005263515.1 | 2 - 3 | 1308 | 79 | |
XM_005263460.4 | XP_005263517.1 | 2 - 3 | 926 | 79 | |
XM_005263462.4 | XP_005263519.1 | 2 - 3 | 359 | 79 | |
XM_011541495.2 | XP_011539797.1 | 2 - 3 | 1305 | 79 | |
XM_011541496.2 | XP_011539798.1 | 2 - 3 | 1308 | 79 | |
XM_017001328.1 | XP_016856817.1 | 2 - 3 | 1308 | 79 | |
GenBank mRNA | AB209113.1 | - | 1 - 2 | 509 | 79 |
AJ237672.1 | - | 1 - 2 | 371 | 79 | |
AK312907.1 | - | 2 - 3 | 332 | 79 | |
BC053509.1 | - | 2 - 3 | 293 | 79 |