The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243.4 | NP_001234.3 | 5 - 6 | 736 | 85 |
NM_001281430.2 | NP_001268359.2 | 4 - 5 | 648 | 85 | |
XM_011542441.2 | XP_011540743.1 | 5 - 6 | 795 | 85 | |
XM_011542443.2 | XP_011540745.1 | 4 - 5 | 813 | 85 | |
GenBank mRNA | BC136400.1 | - | 4 - 5 | 555 | 85 |
BC171762.1 | - | 4 - 5 | 555 | 85 | |
M83554.1 | - | 5 - 6 | 736 | 85 | |
S75768.1 | - | 5 - 6 | 514 | 85 |