Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMLN
Entrez Gene ID
89782
Gene Name
leishmanolysin like peptidase
Gene Aliases
GP63, INV, IX14, MSP
Chromosome Location
Chr.3: 197960200 - 198043720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136049.2 NP_001129521.2 16 - 17 2002 67
NM_033029.3 NP_149018.2 15 - 16 1891 67
NR_026786.1 16 - 17 1881 67
NR_026787.1 15 - 16 1770 67
XM_011513270.1 XP_011511572.1 16 - 17 1970 67
XM_017007428.1 XP_016862917.1 15 - 16 1859 67
GenBank mRNA AJ312398.1 - 16 - 17 1881 67
AJ312399.1 - 15 - 16 1891 67
AK299119.1 - 16 - 17 1969 67
AL832783.1 - 15 - 16 2475 67
AM920777.1 - 16 - 17 1980 67
AM920778.1 - 15 - 16 1713 67

View Full Product Details