Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZBTB17
Entrez Gene ID
7709
Gene Name
zinc finger and BTB domain containing 17
Gene Aliases
MIZ-1, ZNF151, ZNF60, pHZ-67
Chromosome Location
Chr.1: 15941869 - 15976132 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242884.1 NP_001229813.1 9 - 10 1513 64
NM_001287603.1 NP_001274532.1 10 - 11 1698 64
NM_001287604.1 NP_001274533.1 10 - 11 1694 64
NM_001324137.1 NP_001311066.1 9 - 10 1491 64
NM_001324138.1 NP_001311067.1 9 - 10 1509 64
NM_003443.2 NP_003434.2 10 - 11 1698 64
XM_005245986.1 XP_005246043.1 9 - 10 1509 64
XM_005245987.1 XP_005246044.1 10 - 11 1639 64
XM_011542085.2 XP_011540387.1 9 - 10 1484 64
XM_011542086.1 XP_011540388.1 10 - 11 1665 64
XM_011542087.2 XP_011540389.1 9 - 10 1506 64
XM_011542088.1 XP_011540390.1 10 - 11 1665 64
XM_017002242.1 XP_016857731.1 8 - 9 1479 64
GenBank mRNA AK223618.1 - 10 - 11 1675 64
AK301896.1 - 9 - 10 1460 64
AK303823.1 - 9 - 10 1404 64
AK315201.1 - 10 - 11 1645 64
BC126163.1 - 9 - 10 1561 64
BC143965.1 - 9 - 10 1561 64
Y09723.1 - 10 - 11 1585 64

View Full Product Details