Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZBTB17
Entrez Gene ID
7709
Gene Name
zinc finger and BTB domain containing 17
Gene Aliases
MIZ-1, ZNF151, ZNF60, pHZ-67
Chromosome Location
Chr.1: 15941869 - 15976132 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242884.1 NP_001229813.1 13 - 14 2093 63
NM_001287604.1 NP_001274533.1 14 - 15 2274 63
NM_001324137.1 NP_001311066.1 13 - 14 2071 63
NM_001324138.1 NP_001311067.1 13 - 14 2089 63
NM_003443.2 NP_003434.2 14 - 15 2278 63
XM_011542088.1 XP_011540390.1 14 - 15 2245 63
GenBank mRNA AK223618.1 - 14 - 15 2255 63
AK226168.1 - 14 - 15 2218 63
AK301896.1 - 13 - 14 2040 63
AK303823.1 - 13 - 14 1984 63
AK315201.1 - 14 - 15 2225 63
BC126163.1 - 13 - 14 2141 63
BQ575263.1 - 2 - 3 63
U20647.1 - 2 - 3 302 63
Y09723.1 - 14 - 15 2165 63

View Full Product Details