The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199201.1 | NP_001186130.1 | 15 - 16 | 1611 | 100 |
NM_001199202.1 | NP_001186131.1 | 15 - 16 | 1559 | 100 | |
NM_024687.3 | NP_078963.2 | 15 - 16 | 1611 | 100 | |
XM_011513144.1 | XP_011511446.1 | 15 - 16 | 1611 | 100 | |
XM_011513145.2 | XP_011511447.2 | 14 - 15 | 1637 | 100 | |
XM_011513148.2 | XP_011511450.2 | 15 - 16 | 1611 | 100 | |
XM_011513149.2 | XP_011511451.2 | 16 - 17 | 1728 | 100 | |
XM_011513151.2 | XP_011511453.2 | 17 - 18 | 1917 | 100 | |
XM_011513152.2 | XP_011511454.2 | 9 - 10 | 1038 | 100 | |
XM_011513153.2 | XP_011511455.2 | 6 - 7 | 710 | 100 | |
XM_017007186.1 | XP_016862675.1 | 15 - 16 | 1611 | 100 | |
XM_017007187.1 | XP_016862676.1 | 15 - 16 | 1611 | 100 | |
XM_017007188.1 | XP_016862677.1 | 15 - 16 | 1611 | 100 | |
XM_017007189.1 | XP_016862678.1 | 14 - 15 | 1637 | 100 | |
XM_017007190.1 | XP_016862679.1 | 15 - 16 | 1611 | 100 | |
GenBank mRNA | AK026702.1 | - | 5 - 6 | 460 | 100 |
AK093296.1 | - | 15 - 16 | 1597 | 100 | |
BC034229.1 | - | 15 - 16 | 1558 | 100 | |
BC144444.1 | - | 14 - 15 | 1558 | 100 |