Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBBX
Entrez Gene ID
79740
Gene Name
zinc finger B-box domain containing
Gene Aliases
-
Chromosome Location
Chr.3: 167178399 - 167380405 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199201.1 NP_001186130.1 18 - 19 2215 102
XM_011513144.1 XP_011511446.1 18 - 19 2215 102
XM_011513145.2 XP_011511447.2 17 - 18 2241 102
XM_011513148.2 XP_011511450.2 18 - 19 2212 102
XM_011513149.2 XP_011511451.2 19 - 20 2332 102
XM_011513151.2 XP_011511453.2 20 - 21 2521 102
XM_011513152.2 XP_011511454.2 12 - 13 1642 102
XM_011513153.2 XP_011511455.2 9 - 10 1314 102
XM_017007186.1 XP_016862675.1 18 - 19 2215 102
XM_017007187.1 XP_016862676.1 18 - 19 2215 102
XM_017007190.1 XP_016862679.1 18 - 19 2215 102
GenBank mRNA AK026702.1 - 9 - 10 1064 102
BC144444.1 - 17 - 18 2162 102

View Full Product Details