Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLCO2A1
Entrez Gene ID
6578
Gene Name
solute carrier organic anion transporter family member 2A1
Gene Aliases
MATR1, OATP2A1, PGT, PHOAR2, SLC21A2
Chromosome Location
Chr.3: 133932696 - 134030076 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005630.2 NP_005621.2 11 - 12 1900 72
XM_017007075.1 XP_016862564.1 11 - 12 1843 72
XM_017007076.1 XP_016862565.1 11 - 12 1843 72
XM_017007077.1 XP_016862566.1 12 - 13 1888 72
GenBank mRNA BC041140.1 - 11 - 12 1729 72
BC051347.1 - 11 - 12 1809 72
U70867.1 - 11 - 12 1709 72

View Full Product Details