Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D5
Entrez Gene ID
9779
Gene Name
TBC1 domain family member 5
Gene Aliases
-
Chromosome Location
Chr.3: 17157162 - 17742739 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134381.1 NP_001127853.1 20 - 21 2083 92
NM_014744.2 NP_055559.1 18 - 19 3418 92
XM_005265611.1 XP_005265668.1 19 - 20 2054 92
XM_005265612.1 XP_005265669.1 18 - 19 1989 92
XM_005265614.1 XP_005265671.1 18 - 19 1988 92
XM_005265615.1 XP_005265672.1 18 - 19 1984 92
XM_005265616.4 XP_005265673.1 17 - 18 1913 92
XM_006713430.1 XP_006713493.1 19 - 20 2104 92
XM_011534281.1 XP_011532583.1 21 - 22 2365 92
XM_011534283.2 XP_011532585.1 20 - 21 2143 92
XM_011534284.2 XP_011532586.1 20 - 21 2188 92
XM_011534286.1 XP_011532588.1 19 - 20 2120 92
XM_011534287.1 XP_011532589.1 19 - 20 2102 92
XM_017007552.1 XP_016863041.1 22 - 23 2466 92
XM_017007553.1 XP_016863042.1 22 - 23 2496 92
XM_017007554.1 XP_016863043.1 27 - 28 3109 92
XM_017007555.1 XP_016863044.1 21 - 22 2348 92
XM_017007556.1 XP_016863045.1 20 - 21 2328 92
XM_017007557.1 XP_016863046.1 17 - 18 1882 92
XM_017007558.1 XP_016863047.1 17 - 18 1912 92
XM_017007559.1 XP_016863048.1 19 - 20 2262 92
XM_017007560.1 XP_016863049.1 19 - 20 2092 92
XM_017007561.1 XP_016863050.1 17 - 18 1923 92
XM_017007562.1 XP_016863051.1 19 - 20 2102 92
XM_017007563.1 XP_016863052.1 16 - 17 1842 92
XM_017007564.1 XP_016863053.1 17 - 18 1918 92
XM_017007565.1 XP_016863054.1 18 - 19 2216 92
XM_017007566.1 XP_016863055.1 18 - 19 2005 92
XM_017007567.1 XP_016863056.1 16 - 17 1846 92
GenBank mRNA AB449886.1 - 16 - 17 1753 92
AK097990.1 - 5 - 6 862 92
AK309374.1 - 15 - 16 1744 92
AK310539.1 - 20 - 21 2083 92
BC013145.1 - 17 - 18 1930 92
D86965.1 - 18 - 19 3417 92

View Full Product Details