The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011534284.2 | XP_011532586.1 | 3 - 4 | 335 | 153 |
XM_011534287.1 | XP_011532589.1 | 3 - 4 | 319 | 153 |