Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D5
Entrez Gene ID
9779
Gene Name
TBC1 domain family member 5
Gene Aliases
-
Chromosome Location
Chr.3: 17157162 - 17742739 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134381.1 NP_001127853.1 5 - 6 428 146
NM_014744.2 NP_055559.1 4 - 5 1829 146
XM_005265611.1 XP_005265668.1 4 - 5 399 146
XM_005265612.1 XP_005265669.1 3 - 4 334 146
XM_005265614.1 XP_005265671.1 4 - 5 399 146
XM_006713430.1 XP_006713493.1 4 - 5 449 146
XM_011534281.1 XP_011532583.1 6 - 7 710 146
XM_011534283.2 XP_011532585.1 5 - 6 488 146
XM_011534284.2 XP_011532586.1 5 - 6 533 146
XM_011534286.1 XP_011532588.1 4 - 5 465 146
XM_017007552.1 XP_016863041.1 7 - 8 811 146
XM_017007553.1 XP_016863042.1 7 - 8 841 146
XM_017007554.1 XP_016863043.1 12 - 13 1454 146
XM_017007555.1 XP_016863044.1 6 - 7 693 146
XM_017007556.1 XP_016863045.1 5 - 6 673 146
XM_017007557.1 XP_016863046.1 3 - 4 293 146
XM_017007558.1 XP_016863047.1 3 - 4 323 146
XM_017007559.1 XP_016863048.1 5 - 6 673 146
XM_017007560.1 XP_016863049.1 5 - 6 503 146
XM_017007561.1 XP_016863050.1 3 - 4 334 146
XM_017007568.1 XP_016863057.1 3 - 4 293 146
GenBank mRNA AB449886.1 - 2 - 3 164 146
AK310539.1 - 5 - 6 428 146
BC013145.1 - 3 - 4 341 146
D86965.1 - 4 - 5 1828 146

View Full Product Details