Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIX1L
Entrez Gene ID
128077
Gene Name
limb and CNS expressed 1 like
Gene Aliases
-
Chromosome Location
Chr.1: 145933423 - 145958038 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153713.2 NP_714924.1 4 - 5 785 75
GenBank mRNA AK123675.1 - 1 - 2 37 75
AK128733.1 - 5 - 6 626 75
BC035727.1 - 4 - 5 767 75
CR627381.1 - 3 - 4 423 75

View Full Product Details