The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001640.3 | NP_001631.3 | 15 - 16 | 1836 | 83 |
XM_005265097.2 | XP_005265154.1 | 14 - 15 | 1706 | 83 | |
XM_005265098.4 | XP_005265155.1 | 14 - 15 | 1375 | 83 | |
XM_011533658.2 | XP_011531960.1 | 15 - 16 | 1583 | 83 | |
XM_011533659.1 | XP_011531961.1 | 15 - 16 | 1535 | 83 | |
XM_011533660.2 | XP_011531962.1 | 16 - 17 | 1724 | 83 | |
XM_011533661.2 | XP_011531963.1 | 14 - 15 | 2572 | 83 | |
XM_011533662.2 | XP_011531964.1 | 13 - 14 | 1190 | 83 | |
XM_011533663.2 | XP_011531965.1 | 12 - 13 | 1035 | 83 | |
XM_017006285.1 | XP_016861774.1 | 14 - 15 | 1445 | 83 | |
XM_017006286.1 | XP_016861775.1 | 13 - 14 | 1189 | 83 | |
GenBank mRNA | AF141383.1 | - | 14 - 15 | 1436 | 83 |
AK314962.1 | - | 15 - 16 | 1471 | 83 | |
BC000362.2 | - | 15 - 16 | 1836 | 83 | |
BC001499.2 | - | 15 - 16 | 1836 | 83 | |
BC001826.2 | - | 15 - 16 | 1836 | 83 | |
D38441.1 | - | 15 - 16 | 1455 | 83 |