The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001640.3 | NP_001631.3 | 18 - 19 | 2092 | 56 |
XM_005265097.2 | XP_005265154.1 | 17 - 18 | 1962 | 56 | |
XM_005265098.4 | XP_005265155.1 | 17 - 18 | 1631 | 56 | |
XM_011533658.2 | XP_011531960.1 | 18 - 19 | 1839 | 56 | |
XM_011533659.1 | XP_011531961.1 | 18 - 19 | 1791 | 56 | |
XM_011533660.2 | XP_011531962.1 | 19 - 20 | 1980 | 56 | |
XM_011533661.2 | XP_011531963.1 | 17 - 18 | 2828 | 56 | |
XM_011533662.2 | XP_011531964.1 | 16 - 17 | 1446 | 56 | |
XM_011533663.2 | XP_011531965.1 | 15 - 16 | 1291 | 56 | |
XM_017006285.1 | XP_016861774.1 | 17 - 18 | 1701 | 56 | |
XM_017006286.1 | XP_016861775.1 | 16 - 17 | 1445 | 56 | |
GenBank mRNA | AF141383.1 | - | 17 - 18 | 1692 | 56 |
AK314962.1 | - | 18 - 19 | 1727 | 56 | |
BC000362.2 | - | 18 - 19 | 2092 | 56 | |
BC001499.2 | - | 18 - 19 | 2092 | 56 | |
BC001826.2 | - | 18 - 19 | 2092 | 56 | |
D38441.1 | - | 18 - 19 | 1711 | 56 |