The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024795.3 | NP_079071.2 | 2 - 3 | 281 | 102 |
XM_011511876.2 | XP_011510178.1 | 2 - 3 | 187 | 102 | |
GenBank mRNA | AK026453.1 | - | 2 - 3 | 280 | 102 |
AY358671.1 | - | 2 - 3 | 273 | 102 | |
BC035754.1 | - | 2 - 3 | 268 | 102 | |
BC137256.1 | - | 2 - 3 | 267 | 102 | |
BC137257.1 | - | 2 - 3 | 267 | 102 | |
DA926038.1 | - | 2 - 3 | 281 | 102 |