The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013379.2 | NP_037511.2 | 7 - 8 | 905 | 86 |
XM_005266075.3 | XP_005266132.1 | 7 - 8 | 1135 | 86 | |
XM_006717083.3 | XP_006717146.1 | 7 - 8 | 1136 | 86 | |
XM_011518599.2 | XP_011516901.1 | 6 - 7 | 956 | 86 | |
XM_011518600.1 | XP_011516902.1 | 7 - 8 | 866 | 86 | |
XM_017014651.1 | XP_016870140.1 | 6 - 7 | 956 | 86 | |
XM_017014652.1 | XP_016870141.1 | 7 - 8 | 916 | 86 | |
GenBank mRNA | AB209787.1 | - | 7 - 8 | 872 | 86 |
AF154502.1 | - | 7 - 8 | 875 | 86 | |
AK292112.1 | - | 7 - 8 | 882 | 86 | |
AK293700.1 | - | 6 - 7 | 870 | 86 | |
AK298288.1 | - | 6 - 7 | 738 | 86 | |
BC011907.2 | - | 7 - 8 | 880 | 86 | |
BC016961.1 | - | 7 - 8 | 880 | 86 |