The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001640.3 | NP_001631.3 | 2 - 3 | 550 | 84 |
XM_005265097.2 | XP_005265154.1 | 2 - 3 | 558 | 84 | |
XM_011533660.2 | XP_011531962.1 | 3 - 4 | 438 | 84 | |
XM_011533661.2 | XP_011531963.1 | 2 - 3 | 1450 | 84 | |
GenBank mRNA | AF141383.1 | - | 1 - 2 | 150 | 84 |
AK314962.1 | - | 2 - 3 | 185 | 84 | |
BC000362.2 | - | 2 - 3 | 550 | 84 | |
BC001499.2 | - | 2 - 3 | 550 | 84 | |
BC001826.2 | - | 2 - 3 | 550 | 84 | |
S90797.1 | - | 1 - 2 | 104 | 84 |