The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006336.3 | NP_006327.2 | 12 - 13 | 2296 | 90 |
XM_005251645.2 | XP_005251702.1 | 12 - 13 | 2088 | 90 | |
XM_005251646.2 | XP_005251703.1 | 12 - 13 | 2285 | 90 | |
XM_011518122.2 | XP_011516424.1 | 12 - 13 | 2098 | 90 | |
XM_017014185.1 | XP_016869674.1 | 12 - 13 | 2704 | 90 | |
XM_017014186.1 | XP_016869675.1 | 12 - 13 | 2079 | 90 | |
XM_017014187.1 | XP_016869676.1 | 12 - 13 | 2698 | 90 | |
XM_017014188.1 | XP_016869677.1 | 12 - 13 | 2066 | 90 | |
GenBank mRNA | AK314911.1 | - | 12 - 13 | 2044 | 90 |
BC001447.2 | - | 1 - 2 | 91 | 90 | |
BC052563.1 | - | 12 - 13 | 2281 | 90 | |
X99802.1 | - | 11 - 12 | 1886 | 90 |