The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006336.3 | NP_006327.2 | 13 - 14 | 2496 | 89 |
XM_005251645.2 | XP_005251702.1 | 13 - 14 | 2288 | 89 | |
XM_005251646.2 | XP_005251703.1 | 13 - 14 | 2485 | 89 | |
XM_011518122.2 | XP_011516424.1 | 13 - 14 | 2298 | 89 | |
XM_017014185.1 | XP_016869674.1 | 13 - 14 | 2904 | 89 | |
XM_017014186.1 | XP_016869675.1 | 13 - 14 | 2279 | 89 | |
XM_017014187.1 | XP_016869676.1 | 13 - 14 | 2898 | 89 | |
XM_017014188.1 | XP_016869677.1 | 13 - 14 | 2266 | 89 | |
GenBank mRNA | AK314911.1 | - | 13 - 14 | 2244 | 89 |
BC001447.2 | - | 2 - 3 | 291 | 89 | |
BC052563.1 | - | 13 - 14 | 2481 | 89 | |
X99802.1 | - | 12 - 13 | 2086 | 89 |