The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006336.3 | NP_006327.2 | 2 - 3 | 612 | 67 |
XM_005251645.2 | XP_005251702.1 | 2 - 3 | 404 | 67 | |
XM_005251646.2 | XP_005251703.1 | 2 - 3 | 607 | 67 | |
XM_011518122.2 | XP_011516424.1 | 2 - 3 | 414 | 67 | |
XM_017014185.1 | XP_016869674.1 | 2 - 3 | 1020 | 67 | |
XM_017014186.1 | XP_016869675.1 | 2 - 3 | 401 | 67 | |
XM_017014187.1 | XP_016869676.1 | 2 - 3 | 1020 | 67 | |
XM_017014188.1 | XP_016869677.1 | 2 - 3 | 388 | 67 | |
XM_017014189.1 | XP_016869678.1 | 2 - 3 | 607 | 67 | |
XM_017014190.1 | XP_016869679.1 | 2 - 3 | 401 | 67 | |
GenBank mRNA | AK314911.1 | - | 2 - 3 | 360 | 67 |
BC033884.1 | - | 2 - 3 | 581 | 67 | |
BC052563.1 | - | 2 - 3 | 597 | 67 | |
X99802.1 | - | 1 - 2 | 202 | 67 |