The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006336.3 | NP_006327.2 | 3 - 4 | 759 | 104 |
XM_005251645.2 | XP_005251702.1 | 3 - 4 | 551 | 104 | |
XM_005251646.2 | XP_005251703.1 | 3 - 4 | 754 | 104 | |
XM_011518122.2 | XP_011516424.1 | 3 - 4 | 561 | 104 | |
XM_017014185.1 | XP_016869674.1 | 3 - 4 | 1167 | 104 | |
XM_017014186.1 | XP_016869675.1 | 3 - 4 | 548 | 104 | |
XM_017014187.1 | XP_016869676.1 | 3 - 4 | 1167 | 104 | |
XM_017014188.1 | XP_016869677.1 | 3 - 4 | 535 | 104 | |
XM_017014189.1 | XP_016869678.1 | 3 - 4 | 754 | 104 | |
XM_017014190.1 | XP_016869679.1 | 3 - 4 | 548 | 104 | |
GenBank mRNA | AK314911.1 | - | 3 - 4 | 507 | 104 |
BC033884.1 | - | 3 - 4 | 728 | 104 | |
BC052563.1 | - | 3 - 4 | 744 | 104 | |
X99802.1 | - | 2 - 3 | 349 | 104 |