The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006336.3 | NP_006327.2 | 7 - 8 | 1632 | 77 |
XM_005251645.2 | XP_005251702.1 | 7 - 8 | 1424 | 77 | |
XM_005251646.2 | XP_005251703.1 | 7 - 8 | 1627 | 77 | |
XM_011518122.2 | XP_011516424.1 | 7 - 8 | 1434 | 77 | |
XM_017014185.1 | XP_016869674.1 | 7 - 8 | 2040 | 77 | |
XM_017014186.1 | XP_016869675.1 | 7 - 8 | 1421 | 77 | |
XM_017014187.1 | XP_016869676.1 | 7 - 8 | 2040 | 77 | |
XM_017014188.1 | XP_016869677.1 | 7 - 8 | 1408 | 77 | |
XM_017014189.1 | XP_016869678.1 | 7 - 8 | 1627 | 77 | |
XM_017014190.1 | XP_016869679.1 | 7 - 8 | 1421 | 77 | |
GenBank mRNA | AK314911.1 | - | 7 - 8 | 1380 | 77 |
BC052563.1 | - | 7 - 8 | 1617 | 77 | |
X99802.1 | - | 6 - 7 | 1222 | 77 |