Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MECOM
Entrez Gene ID
2122
Gene Name
MDS1 and EVI1 complex locus
Gene Aliases
AML1-EVI-1, EVI1, MDS1, MDS1-EVI1, PRDM3, RUSAT2
Chromosome Location
Chr.3: 169083499 - 169663781 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105077.3 NP_001098547.3 7 - 8 900 79
NM_001105078.3 NP_001098548.2 6 - 7 891 79
NM_001163999.1 NP_001157471.1 6 - 7 838 79
NM_001164000.1 NP_001157472.1 6 - 7 1767 79
NM_001205194.1 NP_001192123.1 6 - 7 1195 79
NM_004991.3 NP_004982.2 7 - 8 1533 79
NM_005241.3 NP_005232.2 6 - 7 835 79
XM_005247213.3 XP_005247270.1 7 - 8 1542 79
XM_005247214.3 XP_005247271.1 7 - 8 1542 79
XM_005247215.3 XP_005247272.1 7 - 8 1538 79
XM_005247219.2 XP_005247276.1 6 - 7 1507 79
XM_005247220.2 XP_005247277.1 6 - 7 848 79
XM_005247221.2 XP_005247278.1 6 - 7 831 79
XM_005247223.2 XP_005247280.1 6 - 7 1504 79
XM_011512546.2 XP_011510848.1 6 - 7 1295 79
XM_017005874.1 XP_016861363.1 6 - 7 2210 79
XM_017005875.1 XP_016861364.1 6 - 7 845 79
XM_017005876.1 XP_016861365.1 6 - 7 1507 79
GenBank mRNA AK292865.1 - 6 - 7 818 79
AK304098.1 - 7 - 8 1479 79
AK310509.1 - 5 - 6 679 79
BC130520.1 - 7 - 8 874 79
BC143951.1 - 7 - 8 874 79
BC143952.1 - 6 - 7 812 79
BX640908.1 - 6 - 7 1767 79
BX647613.1 - 6 - 7 1499 79
GQ352634.1 - 6 - 7 1099 79
X54989.1 - 6 - 7 833 79

View Full Product Details