Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RALGPS1
Entrez Gene ID
9649
Gene Name
Ral GEF with PH domain and SH3 binding motif 1
Gene Aliases
RALGEF2, RALGPS1A
Chromosome Location
Chr.9: 126914774 - 127223166 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190728.1 NP_001177657.1 9 - 10 905 115
NM_001190729.1 NP_001177658.1 10 - 11 1144 115
NM_001322320.1 NP_001309249.1 10 - 11 1107 115
NM_001322321.1 NP_001309250.1 10 - 11 1144 115
NM_001322322.1 NP_001309251.1 10 - 11 1144 115
NM_001322323.1 NP_001309252.1 9 - 10 1054 115
NM_001322324.1 NP_001309253.1 10 - 11 1144 115
NM_001322325.1 NP_001309254.1 10 - 11 1144 115
NM_014636.2 NP_055451.1 10 - 11 1107 115
XM_006717328.3 XP_006717391.2 11 - 12 1118 115
XM_011519225.1 XP_011517527.1 10 - 11 1067 115
XM_011519226.1 XP_011517528.1 10 - 11 1067 115
XM_011519227.1 XP_011517529.1 9 - 10 1016 115
XM_011519228.1 XP_011517530.1 10 - 11 1067 115
XM_011519229.1 XP_011517531.1 11 - 12 1124 115
XM_011519230.2 XP_011517532.1 11 - 12 1118 115
XM_011519231.1 XP_011517533.1 10 - 11 1067 115
XM_011519232.1 XP_011517534.1 10 - 11 1067 115
XM_011519233.1 XP_011517535.1 9 - 10 905 115
XM_011519234.2 XP_011517536.1 10 - 11 1201 115
XM_011519235.2 XP_011517537.1 11 - 12 1292 115
XM_011519236.1 XP_011517538.1 9 - 10 905 115
XM_011519238.2 XP_011517540.1 10 - 11 1067 115
XM_017015342.1 XP_016870831.1 10 - 11 1067 115
XM_017015343.1 XP_016870832.1 11 - 12 1111 115
XM_017015344.1 XP_016870833.1 8 - 9 884 115
XM_017015345.1 XP_016870834.1 10 - 11 1067 115
XM_017015346.1 XP_016870835.1 9 - 10 905 115
XM_017015347.1 XP_016870836.1 10 - 11 964 115
XM_017015348.1 XP_016870837.1 10 - 11 1138 115
XM_017015349.1 XP_016870838.1 7 - 8 752 115
GenBank mRNA AB002349.1 - 10 - 11 1107 115
AF221098.1 - 9 - 10 905 115
AK299149.1 - 10 - 11 1085 115
BX648170.1 - 3 - 4 1002 115

View Full Product Details