Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RALGPS1
Entrez Gene ID
9649
Gene Name
Ral GEF with PH domain and SH3 binding motif 1
Gene Aliases
RALGEF2, RALGPS1A
Chromosome Location
Chr.9: 126914774 - 127223166 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322321.1 NP_001309250.1 13 - 14 1498 79
NM_001322322.1 NP_001309251.1 13 - 14 1498 79
NM_001322325.1 NP_001309254.1 13 - 14 1498 79
NM_014636.2 NP_055451.1 13 - 14 1461 79
XM_006717328.3 XP_006717391.2 14 - 15 1472 79
XM_011519225.1 XP_011517527.1 13 - 14 1421 79
XM_011519226.1 XP_011517528.1 13 - 14 1421 79
XM_011519227.1 XP_011517529.1 12 - 13 1370 79
XM_011519228.1 XP_011517530.1 13 - 14 1421 79
XM_011519229.1 XP_011517531.1 14 - 15 1478 79
XM_011519230.2 XP_011517532.1 14 - 15 1472 79
XM_011519233.1 XP_011517535.1 12 - 13 1259 79
XM_011519234.2 XP_011517536.1 13 - 14 1555 79
XM_011519235.2 XP_011517537.1 14 - 15 1646 79
XM_011519238.2 XP_011517540.1 13 - 14 1421 79
XM_017015342.1 XP_016870831.1 13 - 14 1421 79
XM_017015343.1 XP_016870832.1 14 - 15 1465 79
XM_017015344.1 XP_016870833.1 11 - 12 1238 79
XM_017015346.1 XP_016870835.1 12 - 13 1259 79
XM_017015347.1 XP_016870836.1 13 - 14 1318 79
XM_017015349.1 XP_016870838.1 10 - 11 1106 79
GenBank mRNA AB002349.1 - 13 - 14 1461 79

View Full Product Details