Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RALGPS1
Entrez Gene ID
9649
Gene Name
Ral GEF with PH domain and SH3 binding motif 1
Gene Aliases
RALGEF2, RALGPS1A
Chromosome Location
Chr.9: 126914774 - 127223166 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190728.1 NP_001177657.1 6 - 7 550 86
NM_001190729.1 NP_001177658.1 7 - 8 789 86
NM_001190730.1 NP_001177659.1 7 - 8 789 86
NM_001322320.1 NP_001309249.1 7 - 8 752 86
NM_001322321.1 NP_001309250.1 7 - 8 789 86
NM_001322322.1 NP_001309251.1 7 - 8 789 86
NM_001322323.1 NP_001309252.1 6 - 7 699 86
NM_001322324.1 NP_001309253.1 7 - 8 789 86
NM_001322325.1 NP_001309254.1 7 - 8 789 86
NM_014636.2 NP_055451.1 7 - 8 752 86
XM_006717328.3 XP_006717391.2 7 - 8 601 86
XM_011519225.1 XP_011517527.1 6 - 7 550 86
XM_011519226.1 XP_011517528.1 6 - 7 550 86
XM_011519227.1 XP_011517529.1 5 - 6 499 86
XM_011519228.1 XP_011517530.1 6 - 7 550 86
XM_011519229.1 XP_011517531.1 7 - 8 607 86
XM_011519230.2 XP_011517532.1 7 - 8 601 86
XM_011519231.1 XP_011517533.1 6 - 7 550 86
XM_011519232.1 XP_011517534.1 6 - 7 550 86
XM_011519233.1 XP_011517535.1 6 - 7 550 86
XM_011519234.2 XP_011517536.1 6 - 7 684 86
XM_011519235.2 XP_011517537.1 7 - 8 775 86
XM_011519236.1 XP_011517538.1 6 - 7 550 86
XM_011519238.2 XP_011517540.1 6 - 7 550 86
XM_017015342.1 XP_016870831.1 6 - 7 550 86
XM_017015343.1 XP_016870832.1 7 - 8 594 86
XM_017015345.1 XP_016870834.1 6 - 7 550 86
XM_017015346.1 XP_016870835.1 6 - 7 550 86
XM_017015347.1 XP_016870836.1 7 - 8 609 86
XM_017015348.1 XP_016870837.1 7 - 8 783 86
XM_017015349.1 XP_016870838.1 3 - 4 235 86
XM_017015350.1 XP_016870839.1 6 - 7 550 86
GenBank mRNA AB002349.1 - 7 - 8 752 86
AF221098.1 - 6 - 7 550 86
AK299149.1 - 7 - 8 730 86
BC032372.1 - 7 - 8 712 86
BC033708.1 - 7 - 8 782 86
DR001330.1 - 7 - 8 744 86

View Full Product Details