The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145099.1 | NP_001138571.1 | 6 - 7 | 1019 | 64 |
NM_017585.3 | NP_060055.2 | 6 - 7 | 1019 | 64 | |
XM_011518189.2 | XP_011516491.2 | 6 - 7 | 996 | 64 | |
XM_017014237.1 | XP_016869726.1 | 6 - 7 | 987 | 64 | |
XM_017014238.1 | XP_016869727.1 | 6 - 7 | 991 | 64 | |
GenBank mRNA | AJ011372.2 | - | 6 - 7 | 961 | 64 |
AK074836.1 | - | 6 - 7 | 969 | 64 | |
AK096630.1 | - | 5 - 6 | 1517 | 64 | |
AK222919.1 | - | 6 - 7 | 964 | 64 | |
AK225106.1 | - | 6 - 7 | 892 | 64 | |
BC013740.1 | - | 6 - 7 | 953 | 64 | |
Y17803.1 | - | 6 - 7 | 970 | 64 |