The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001932.4 | NP_001923.2 | 17 - 18 | 1521 | 79 |
NR_003562.2 | 16 - 17 | 1500 | 79 | ||
XM_006721915.2 | XP_006721978.1 | 18 - 19 | 1709 | 79 | |
XM_011524828.2 | XP_011523130.1 | 17 - 18 | 1604 | 79 | |
XM_017024656.1 | XP_016880145.1 | 17 - 18 | 1737 | 79 | |
XM_017024657.1 | XP_016880146.1 | 16 - 17 | 1413 | 79 | |
GenBank mRNA | AK313045.1 | - | 17 - 18 | 1488 | 79 |
AM050144.1 | - | 15 - 16 | 1352 | 79 | |
AM050145.1 | - | 14 - 15 | 1331 | 79 | |
BC035833.1 | - | 16 - 17 | 1473 | 79 | |
BC047017.1 | - | 16 - 17 | 1491 | 79 | |
BC056865.1 | - | 16 - 17 | 1435 | 79 | |
U37707.1 | - | 15 - 16 | 1682 | 79 |