Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MPP3
Entrez Gene ID
4356
Gene Name
membrane palmitoylated protein 3
Gene Aliases
DLG3
Chromosome Location
Chr.17: 43800799 - 43833192 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001932.4 NP_001923.2 18 - 19 1634 148
NR_003562.2 17 - 18 1613 148
XM_006721915.2 XP_006721978.1 19 - 20 1822 148
XM_011524828.2 XP_011523130.1 18 - 19 1717 148
XM_017024656.1 XP_016880145.1 18 - 19 1850 148
XM_017024657.1 XP_016880146.1 17 - 18 1526 148
GenBank mRNA AB209232.1 - 1 - 2 3856 148
AK311368.1 - 16 - 17 1710 148
AK313045.1 - 18 - 19 1601 148
AM050144.1 - 16 - 17 1465 148
AM050145.1 - 15 - 16 1444 148
BC035833.1 - 17 - 18 1586 148
BC047017.1 - 17 - 18 1604 148
BC056865.1 - 17 - 18 1548 148
U37707.1 - 16 - 17 1795 148

View Full Product Details