The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001932.4 | NP_001923.2 | 7 - 8 | 622 | 74 |
NR_003562.2 | 7 - 8 | 622 | 74 | ||
XM_006721915.2 | XP_006721978.1 | 8 - 9 | 810 | 74 | |
XM_006721916.3 | XP_006721979.1 | 7 - 8 | 705 | 74 | |
XM_011524828.2 | XP_011523130.1 | 7 - 8 | 705 | 74 | |
XM_017024655.1 | XP_016880144.1 | 7 - 8 | 705 | 74 | |
XM_017024656.1 | XP_016880145.1 | 7 - 8 | 705 | 74 | |
XM_017024657.1 | XP_016880146.1 | 7 - 8 | 600 | 74 | |
XM_017024658.1 | XP_016880147.1 | 7 - 8 | 705 | 74 | |
XM_017024659.1 | XP_016880148.1 | 7 - 8 | 705 | 74 | |
XM_017024660.1 | XP_016880149.1 | 7 - 8 | 705 | 74 | |
GenBank mRNA | AK299535.1 | - | 7 - 8 | 683 | 74 |
AK309045.1 | - | 8 - 9 | 762 | 74 | |
AK313045.1 | - | 7 - 8 | 589 | 74 | |
AM050144.1 | - | 5 - 6 | 453 | 74 | |
AM050145.1 | - | 5 - 6 | 453 | 74 | |
BC035833.1 | - | 7 - 8 | 595 | 74 | |
BC047017.1 | - | 7 - 8 | 613 | 74 | |
BC056865.1 | - | 6 - 7 | 536 | 74 | |
U37707.1 | - | 5 - 6 | 783 | 74 |